Anti-CST3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80040-RP01-100 80040-RP01-200 80040-RP01-400
103661-070EA 562.97 USD
103661-070 103661-066 103661-068
Anti-CST3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Cystatin C/CST3 (Catalog#80040-R08H; P14841-1; Met1-Ala140). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CSTC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR