Anti-CSTC Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological

80040-R001-50 80040-R001-100
103596-698EA 128.15 USD
103596-698 103661-062
Anti-CSTC Rabbit Monoclonal Antibody [clone: 001]
Antibodies
Anti-Cystatin C Antibody

Type: Primary
Antigen: CSTC
Clonality: Monoclonal
Clone: Clone: 001
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR