Anti-CXCL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80039-RP01-100 80039-RP01-400 80039-RP01-200
103661-060EA 562.97 USD
103661-060 103661-056 103661-058
Anti-CXCL1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CXCL1/MGSA/NAP-3 (Catalog#80039-RNCE; NP_110472.1; Ala25-Lys96). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CXCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR