Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CXCL1 Mouse Monoclonal Antibody [clone: 12]

Supplier: Sino Biological

80039-MM12-200 80039-MM12-50 80039-MM12-100
103661-054EA 214.31 USD InStock
103661-054 103596-692 103661-052
Anti-CXCL1 Mouse Monoclonal Antibody [clone: 12]
Antibodies
Anti-CXCL1 Antibody

Type: Primary
Antigen: CXCL1
Clonality: Monoclonal
Clone: Clone: 12
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty