Anti-SELE Rabbit Polyclonal Antibody

Supplier: Sino Biological

80033-T16-100 80033-T16-50 80033-T16-200
103596-688EA 160.46 USD
103596-688 103589-686 103596-690
Anti-SELE Rabbit Polyclonal Antibody
Antibodies
Anti-E-Selectin Antibody

Type: Primary
Antigen: SELE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR