Anti-SELE Rabbit Monoclonal Antibody [clone: 102]

Supplier: Sino Biological
80033-R102-1 80033-R102-50 80033-R102-100
103589-684EA 1500.59 USD
103589-684 103660-992 103596-684
Anti-SELE Rabbit Monoclonal Antibody [clone: 102]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat E-Selectin / CD62e / SELE (rR E-Selectin / CD62e / SELE; Catalog#80033-R08H; P98105-1; Met1-Pro494).

Type: Primary
Antigen: SELE
Clonality: Monoclonal
Clone: 102
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR