Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SELE Rabbit Monoclonal Antibody [clone: 046]

Supplier: Sino Biological

80033-R046-100 80033-R046-50 80033-R046-1
103660-990EA 214.31 USD InStock
103660-990 103596-682 103589-682
Anti-SELE Rabbit Monoclonal Antibody [clone: 046]
Antibodies
Anti-E-Selectin Antibody

Type: Primary
Antigen: SELE
Clonality: Monoclonal
Clone: Clone: 046
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty