Anti-ACE2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80031-RP01-200 80031-RP01-100 80031-RP01-400
103660-976EA 562.97 USD
103660-976 103660-974 103660-972
Anti-ACE2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat ACE2 (rR ACE2; Catalog#80031-R08H; Q5EGZ1; Met 1-Thr 740). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ACE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR