Anti-ACE2 Rabbit Monoclonal Antibody [clone: 003]

Supplier: Sino Biological
80031-R003-50 80031-R003-100 80031-R003-1
103589-680EA 1500.59 USD
103589-680 103660-968 103596-672
Anti-ACE2 Rabbit Monoclonal Antibody [clone: 003]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ACE2 / Angiotensin-Converting Enzyme 2 (rR ACE2 / Angiotensin-Converting Enzyme 2; Catalog#80031-R08H; Q5EGZ1; Met1-Thr740).

Type: Primary
Antigen: ACE2
Clonality: Monoclonal
Clone: 003
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR