Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-gp130 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80030-T16-200 80030-T16-100 80030-T16-50
103596-666EA 214.31 USD InStock
103596-666 103596-664 103589-676
Anti-gp130 Rabbit Polyclonal Antibody
Antibodies
Anti-gp130 Antibody

Type: Primary
Antigen: gp130
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty