Anti-gp130 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80030-RP01-100 80030-RP01-400 80030-RP01-200
103660-956EA 106.62 USD
103660-956 103660-960 103660-958
Anti-gp130 Rabbit Polyclonal Antibody
Antibodies
Anti-gp130 Antibody

Type: Primary
Antigen: gp130
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR