Anti-gp130 Rabbit Monoclonal Antibody [clone: 108]

Supplier: Sino Biological

80030-R108-50 80030-R108-100
103596-662EA 128.15 USD
103596-662 103660-954
Anti-gp130 Rabbit Monoclonal Antibody [clone: 108]
Antibodies
Anti-gp130 Antibody

Type: Primary
Antigen: gp130
Clonality: Monoclonal
Clone: Clone: 108
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR