Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-gp130 Mouse Monoclonal Antibody [clone: 9D7A9]

Supplier: Sino Biological

80030-MM04-200 80030-MM04-100 80030-MM04-50
103660-952EA 214.31 USD InStock
103660-952 103660-950 103596-660
Anti-gp130 Mouse Monoclonal Antibody [clone: 9D7A9]
Antibodies
Anti-gp130 Antibody

Type: Primary
Antigen: gp130
Clonality: Monoclonal
Clone: Clone: 9D7A9
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty