Anti-IL1R1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80028-RP02-50 80028-RP02-100 80028-RP02-200
103596-656EA 106.62 USD
103596-656 103660-936 103660-938
Anti-IL1R1 Rabbit Polyclonal Antibody
Antibodies
Anti-IL1R1 Antibody

Type: Primary
Antigen: IL1R1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR