Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 027]

Supplier: Sino Biological

80028-R027-100 80028-R027-50
103662-566EA 214.31 USD
103662-566 103596-654
Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 027]
Antibodies
Anti-IL1R1 Antibody

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: Clone: 027
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR