Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 007] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
80028-R007-F-25 80028-R007-F-100
103632-398EA 181.8 USD
103632-398 103630-218
Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 007] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-1R1 / CD121a (rR IL-1R1 / CD121a; Catalog#80028-R08H; NP_037255.3; Met1-Lys352) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: Clone: 007
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR