Anti-IL1R1 Mouse Monoclonal Antibody [clone: 10]

Supplier: Sino Biological
80028-MM10-50 80028-MM10-200 80028-MM10-100 80028-MM10-1
103662-562EA 352.03 USD
103662-562 103661-114 103589-674 103596-650
Anti-IL1R1 Mouse Monoclonal Antibody [clone: 10]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL1R1 / IL-1R1 / IL-1 RI (rR IL1R1 / IL-1R1 / IL-1 RI; Catalog#80028-R08H; NP_037255.3; Met1-Lys352). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: 10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR