Anti-CD86 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80025-RP02-50 80025-RP02-200 80025-RP02-100
103596-642EA 142.96 USD
103596-642 103660-996 103660-994
Anti-CD86 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD86/B7-2 (rR CD86/B7-2; Catalog#80025-R08H; NP_064466.1; Met 1-Ile 250). CD86/B7-2 specific IgG was purified by Rat CD86/B7-2 affinity chromatography.

Type: Primary
Antigen: CD86
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR