Anti-CD86 Mouse Monoclonal Antibody [clone: 12]

Supplier: Sino Biological

80025-MM12-200 80025-MM12-1 80025-MM12-50 80025-MM12-100
103660-930EA 214.31 USD
103660-930 103589-668 103596-640 103660-928
Anti-CD86 Mouse Monoclonal Antibody [clone: 12]
Antibodies
Anti-CD86 Antibody

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: Clone: 12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR