Anti-CD86 Mouse Monoclonal Antibody [clone: 10]

Supplier: Sino Biological

80025-MM10-100 80025-MM10-200 80025-MM10-1 80025-MM10-50
103660-924EA 160.46 USD
103660-924 103660-926 103589-666 103596-638
Anti-CD86 Mouse Monoclonal Antibody [clone: 10]
Antibodies
Anti-CD86 Antibody

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: Clone: 10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR