Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD86 Mouse Monoclonal Antibody [clone: 6B4A7]

Supplier: Sino Biological

80025-MM04-200 80025-MM04-50 80025-MM04-100
103660-922EA 214.31 USD InStock
103660-922 103596-636 103660-920
Anti-CD86 Mouse Monoclonal Antibody [clone: 6B4A7]
Antibodies
Anti-CD86 Antibody

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: Clone: 6B4A7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty