Anti-IL1B Rabbit Polyclonal Antibody

Supplier: Sino Biological
80023-RP01-400 80023-RP01-200 80023-RP01-100
103660-914EA 562.97 USD
103660-914 103660-912 103660-910
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat IL-1 beta (Catalog#80023-RNAE; Q63264-1; Val117-Ser268). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL-1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR