Anti-IL1B Rabbit Monoclonal Antibody [clone: 017]

Supplier: Sino Biological
80023-R017-50 80023-R017-100
103596-628EA 150.18 USD
103596-628 103660-908
Anti-IL1B Rabbit Monoclonal Antibody [clone: 017]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-1 beta (Catalog#80023-RNAE; Q63264-1; Val117-Ser268).

Type: Primary
Antigen: IL-1B
Clonality: Monoclonal
Clone: 017
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR