Anti-ICAM-1 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 037]

Supplier: Sino Biological

80022-R037-P-100 80022-R037-P-25
103632-396EA 96.92 USD
103632-396 103630-216
Anti-ICAM-1 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 037]
Antibodies
Anti-ICAM-1 Antibody (PE)

Type: Primary
Antigen: ICAM-1
Clonality: Monoclonal
Clone: Clone: 037
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR