Anti-ICAM1 Rabbit Monoclonal Antibody [clone: 037] APC (Allophycocyanin)

Supplier: Sino Biological
80022-R037-A-25 80022-R037-A-100
103630-212EA 70.7 USD
103630-212 103631-096
Anti-ICAM1 Rabbit Monoclonal Antibody [clone: 037] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ICAM-1 / CD54 (rR ICAM-1 / CD54; Catalog#80022-R08H; Q00238-1; Met1-Thr493) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: ICAM-1
Clonality: Monoclonal
Clone: Clone: 037
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR