Anti-ICAM1 Rabbit Monoclonal Antibody [clone: 037]

Supplier: Sino Biological
80022-R037-50 80022-R037-1 80022-R037-100
103596-620EA 150.18 USD
103596-620 103588-732 103660-892
Anti-ICAM1 Rabbit Monoclonal Antibody [clone: 037]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ICAM-1 / CD54 (rR ICAM-1 / CD54; Catalog#80022-R08H; Q00238-1; Met1-Thr493).

Type: Primary
Antigen: ICAM-1
Clonality: Monoclonal
Clone: 037
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR