Anti-GFRA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80021-RP02-200 80021-RP02-100 80021-RP02-50
103596-610EA 142.96 USD
103596-610 103660-884 103660-882
Anti-GFRA1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat GFRA1 (rR GFRA1; Catalog#80021-R08H; Q62997-1; Met 1-Leu 445). GFRA1 specific IgG was purified by Rat GFRA1 affinity chromatography.

Type: Primary
Antigen: GFRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR