Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFRA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80021-RP01-100 80021-RP01-200 80021-RP01-400
103660-876EA 106.62 USD InStock
103660-876 103660-878 103660-880
Anti-GFRA1 Rabbit Polyclonal Antibody
Antibodies
Anti-GFR alpha-1 Antibody

Type: Primary
Antigen: GFRA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty