Anti-GFRA1 Rabbit Monoclonal Antibody [clone: 029]

Supplier: Sino Biological
80021-R029-100 80021-R029-50
103596-608EA 150.18 USD
103596-608 103660-874
Anti-GFRA1 Rabbit Monoclonal Antibody [clone: 029]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat GFRA1 (rR GFRA1; Catalog#80021-R08H; Q62997-1; Met1-Leu445).

Type: Primary
Antigen: GFRA1
Clonality: Monoclonal
Clone: 029
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR