Anti-CNTFR Rabbit Polyclonal Antibody

Supplier: Sino Biological
80019-RP02-50 80019-RP02-200 80019-RP02-100
103596-602EA 142.96 USD
103596-602 103660-528 103660-526
Anti-CNTFR Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CNTFR (rR CNTFR; Catalog#80019-R08B; Q08406-1; Met 1-Pro 346). CNTFR specific IgG was purified by Rat CNTFR affinity chromatography.

Type: Primary
Antigen: CNTFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR