Anti-CNTFR Rabbit Polyclonal Antibody

Supplier: Sino Biological

80019-RP01-100 80019-RP01-400 80019-RP01-200
103660-522EA 106.62 USD
103660-522 103660-524 103660-520
Anti-CNTFR Rabbit Polyclonal Antibody
Antibodies
Anti-CNTFR Antibody

Type: Primary
Antigen: CNTFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR