Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNTFR Mouse Monoclonal Antibody [clone: 3C11G4G4]

Supplier: Sino Biological

80019-MM06-100 80019-MM06-200 80019-MM06-50
103660-516EA 160.46 USD InStock
103660-516 103660-518 103596-600
Anti-CNTFR Mouse Monoclonal Antibody [clone: 3C11G4G4]
Antibodies
Anti-CNTFR Antibody

Type: Primary
Antigen: CNTFR
Clonality: Monoclonal
Clone: Clone: 3C11G4G4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty