Anti-FCGR2B Rabbit Polyclonal Antibody

Supplier: Sino Biological
80018-RP02-100 80018-RP02-50 80018-RP02-200
103660-514EA 352.03 USD
103660-514 103596-598 103660-512
Anti-FCGR2B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD32b / Fc gamma RIIB (rR CD32b / Fc gamma RIIB; Catalog#80018-R08H; Q63203-1; Met 1-Pro 212). CD32b / Fc gamma RIIB specific IgG was purified by Rat CD32b / Fc gamma RIIB affinity chromatography.

Type: Primary
Antigen: Anti-CD32B Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR