Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Anti-CD32B Antibody Rabbit Polyclonal Antibody

Supplier: Sino Biological

80018-RP01-200 80018-RP01-400 80018-RP01-100
103660-508EA 160.46 USD InStock
103660-508 103660-510 103660-506
Anti-Anti-CD32B Antibody Rabbit Polyclonal Antibody
Antibodies
Anti-CD32B Antibody

Type: Primary
Antigen: Anti-CD32B Antibody
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty