Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD16a Rabbit Monoclonal Antibody [clone: 041]

Supplier: Sino Biological

80017-R041-100 80017-R041-50
103660-494EA 214.31 USD InStock
103660-494 103596-594
Anti-CD16a Rabbit Monoclonal Antibody [clone: 041]
Antibodies
Anti-CD16a Antibody

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: Clone: 041
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty