Anti-FCGR3A Mouse Monoclonal Antibody [clone: 4B1H6]

Supplier: Sino Biological
80017-MM06-50 80017-MM06-100 80017-MM06-200
103596-592EA 142.96 USD
103596-592 103659-834 103659-836
Anti-FCGR3A Mouse Monoclonal Antibody [clone: 4B1H6]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD16a/FCGR3A (rR CD16a/FCGR3A; Catalog #80017-R08H; Met 1-Pro 200; ABO70343.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 4B1H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR