Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD64 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80016-RP02-200 80016-RP02-50 80016-RP02-100
103659-832EA 214.31 USD InStock
103659-832 103596-586 103659-830
Anti-CD64 Rabbit Polyclonal Antibody
Antibodies
Anti-CD64 Antibody

Type: Primary
Antigen: CD64
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty