Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD64 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 005]

Supplier: Sino Biological

80016-R005-P-100 80016-R005-P-25
103631-094EA 215.38 USD InStock
103631-094 103631-530
Anti-CD64 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 005]
Antibodies
Anti-CD64 Antibody (PE)

Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: Clone: 005
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty