Anti-FCAR Rabbit Polyclonal Antibody

Supplier: Sino Biological
80014-T16-200 80014-T16-100 80014-T16-50
103594-698EA 352.03 USD
103594-698 103594-696 103588-724
Anti-FCAR Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD89/FCAR (rR CD89/FCAR; Catalog#80014-R08H; NP_973721.1; Met1-Asn228). CD89/FCAR specific IgG was purified by Rat CD89/FCAR affinity chromatography.

Type: Primary
Antigen: CD89
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR