Anti-CD89 Rabbit Monoclonal Antibody [clone: 013]

Supplier: Sino Biological

80014-R013-50 80014-R013-100
103594-692EA 128.15 USD
103594-692 103660-468
Anti-CD89 Rabbit Monoclonal Antibody [clone: 013]
Antibodies
Anti-CD89 Antibody

Type: Primary
Antigen: CD89
Clonality: Monoclonal
Clone: Clone: 013
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR