Anti-DLL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80013-T16-50 80013-T16-200 80013-T16-100
103588-722EA 106.62 USD
103588-722 103596-578 103596-576
Anti-DLL1 Rabbit Polyclonal Antibody
Antibodies
Anti-DLL1 Antibody

Type: Primary
Antigen: DLL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR