Anti-DLL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80013-RP02-200 80013-RP02-100 80013-RP02-50
103660-466EA 352.03 USD
103660-466 103660-464 103596-574
Anti-DLL1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat DLL1 (rR DLL1; Catalog#80013-R08H; EDL99825.1; Met 1-Ser 534). DLL1 specific IgG was purified by Rat DLL1 affinity chromatography.

Type: Primary
Antigen: DLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR