Anti-DLL1 Rabbit Monoclonal Antibody [clone: 006]

Supplier: Sino Biological
80013-R006-100 80013-R006-50
103660-456EA 271.4 USD
103660-456 103596-572
Anti-DLL1 Rabbit Monoclonal Antibody [clone: 006]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat DLL1 (rR DLL1; Catalog#80013-R08H; EDL99825.1; Met 1-Ser 534).

Type: Primary
Antigen: DLL1
Clonality: Monoclonal
Clone: 006
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR