Anti-CNTF Rabbit Monoclonal Antibody [clone: 121]

Supplier: Sino Biological

80012-R121-100 80012-R121-50
103660-444EA 214.31 USD
103660-444 103596-568
Anti-CNTF Rabbit Monoclonal Antibody [clone: 121]
Antibodies
Anti-CNTF Antibody

Type: Primary
Antigen: CNTF
Clonality: Monoclonal
Clone: Clone: 121
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR