Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNTF Mouse Monoclonal Antibody [clone: 10]

Supplier: Sino Biological

80012-MM10-50 80012-MM10-100 80012-MM10-200
103596-566EA 106.62 USD InStock
103596-566 103660-872 103660-442
Anti-CNTF Mouse Monoclonal Antibody [clone: 10]
Antibodies
Anti-CNTF Antibody

Type: Primary
Antigen: CNTF
Clonality: Monoclonal
Clone: Clone: 10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG3
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty