Anti-SOST Rabbit Polyclonal Antibody

Supplier: Sino Biological
80009-T16-200 80009-T16-50 80009-T16-100
103589-352EA 142.96 USD
103589-352 103596-564 103596-562
Anti-SOST Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Sclerostin / SOST (rR Sclerostin / SOST; Catalog#80009-R08H; NP_085073.1; Met1-Tyr213). Sclerostin / SOST specific IgG was purified by Rat Sclerostin / SOST affinity chromatography.

Type: Primary
Antigen: SOST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR