Anti-SOST Rabbit Polyclonal Antibody

Supplier: Sino Biological

80009-RP01-400 80009-RP01-200 80009-RP01-100
103660-870EA 214.31 USD
103660-870 103660-866 103660-868
Anti-SOST Rabbit Polyclonal Antibody
Antibodies
Anti-Sclerostin Antibody

Type: Primary
Antigen: SOST
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR