Anti-SOST Rabbit Monoclonal Antibody [clone: 118]

Supplier: Sino Biological

80009-R118-50 80009-R118-100
103596-560EA 128.15 USD
103596-560 103660-440
Anti-SOST Rabbit Monoclonal Antibody [clone: 118]
Antibodies
Anti-Sclerostin Antibody

Type: Primary
Antigen: SOST
Clonality: Monoclonal
Clone: Clone: 118
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR