Anti-RBP4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80008-RP02-100 80008-RP02-200 80008-RP02-50
103596-558EA 142.96 USD
103596-558 103660-438 103660-436
Anti-RBP4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat RBP4 (rR RBP4; Catalog#80008-R08H; NP_037294.1; Met 1-Leu 201). RBP4 specific IgG was purified by Rat RBP4 affinity chromatography.

Type: Primary
Antigen: RBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR