Anti-RBP4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80008-RP01-200 80008-RP01-100 80008-RP01-400
103660-434EA 562.97 USD
103660-434 103660-432 103660-430
Anti-RBP4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat RBP4 (Catalog#80008-R08H; NP_037294.1; Met1-Leu201). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: RBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR